Sodium 2-amino-9-((2-hydroxyethoxy)methyl)-9H-purin-6-olate ≥95%

Supplier: Matrix Scientific

Synonyms: Acyclovir sodium, Sodium 2-((2-amino-6-oxo-1H-purin-9(6H)-yl)methoxy)ethanolate

076125-1G 076125-5G 076125-10G
102544-608EA 41.54 USD
102544-608 102544-610 102544-612
Sodium 2-amino-9-((2-hydroxyethoxy)methyl)-9H-purin-6-olate ≥95%
Sodium 2-amino-9-((2-hydroxyethoxy)methyl)-9H-purin-6-olate

Hazard: Irritant

Formula: C₈H₁₀N₅NaO₃
MW: 247.19 g/mol
MDL Number: MFCD01694138
CAS Number: 69657-51-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR