Phenylhydrazinium chloride ≥95%

Supplier: Matrix Scientific

Synonyms: Phenylhydrazine hydrochloride, Phenylhydrazinium chloride

076124-10G 076124-100G
102544-604EA 23.08 USD
102544-604 102544-606
Phenylhydrazinium chloride ≥95%
Phenylhydrazinium chloride

Hazard: Irritant

Formula: C₆H₉ClN₂
MDL Number: MFCD00012924
CAS Number: 59-88-1
UN: 2811
ADR: 6.1,III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=e16d43c3611e20636f31c95d3974860d83c8913f&vid=c26f549d3aed1b69060ac1042fd1114ca4edb2cb&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR