1,3-Bis(diphenylphosphino)propane ≥95%

Supplier: Matrix Scientific

Synonyms: Propylenebis(diphenylphosphine), Propane-1,3-diylbis(diphenylphosphane), Propane-1,3-diylbis(diphenylphosphine), 1,3-Bis(diphenylphosphino)propane, DPPP, Bis(1,2-diphenylphosphino)propane

076123-100G 076123-25G
102544-602EA 260 USD
102544-602 102552-016
1,3-Bis(diphenylphosphino)propane ≥95%
1,3-Bis(diphenylphosphino)propane

Hazard: Irritant/Air Sensitive

Formula: C₂₇H₂₆P₂
Melting Pt: 58…63 °C
MDL Number: MFCD00003050
CAS Number: 6737-42-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR