2,3-Dichlorobenzaldehyde ≥95%

Supplier: Matrix Scientific

076122-10G 076122-100G
102544-596EA 36.92 USD
102544-596 102544-598
2,3-Dichlorobenzaldehyde ≥95%
2,3-Dichlorobenzaldehyde

Hazard: Irritant

Formula: C₇H₄Cl₂O
MDL Number: MFCD00010127
CAS Number: 6334-18-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR