2-((2-(2-Hydroxyethoxy)-4-nitrophenyl)amino)ethanol ≥95%

Supplier: Matrix Scientific

076120-1G 076120-5G 076120-10G
102544-586EA 90.77 USD
102544-586 102544-588 102544-590
2-((2-(2-Hydroxyethoxy)-4-nitrophenyl)amino)ethanol ≥95%
2-((2-(2-Hydroxyethoxy)-4-nitrophenyl)amino)ethanol

Hazard: Irritant

Formula: C₁₀H₁₄N₂O₅
MDL Number: MFCD03265655
CAS Number: 59820-43-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR