Cephalothin ≥95%

Supplier: Matrix Scientific

Synonyms: (6R,7R)-3-(Acetoxymethyl)-8-oxo-7-(2-(thiophen-2-yl)acetamido)-5-thia-1-azabicyclo[4.2.0]oct-2-ene-2-carboxylic acid, 7-(2-Thienylacetamido)cephalosporanic Acid, Cephalotin

076118-1G 076118-5G
102544-576EA 38.46 USD
102544-576 102544-578
Cephalothin ≥95%
Cephalothin

Hazard: Irritant

Formula: C₁₆H₁₆N₂O₆S₂
MDL Number: MFCD00242614
CAS Number: 153-61-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR