Dimethyl-4-nitrophthalate ≥95%

Supplier: Matrix Scientific

076115-5G 076115-10G 076115-100G
102544-562EA 143.08 USD
102544-562 102544-564 102544-566
Dimethyl-4-nitrophthalate ≥95%
Dimethyl 4-nitrophthalate

Hazard: Irritant

Formula: C₁₀H₉NO₆
MDL Number: MFCD00017191
CAS Number: 610-22-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR