2-(3-Trifluoromethylanilino)nicotinic acid ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[(3-Trifluoromethylphenyl)amino]nicotinic Acid, 2-((3-(Trifluoromethyl)phenyl)amino)nicotinic acid, Niflumic Acid

076113-10G 076113-100G
102544-554EA 72.31 USD
102544-554 102544-556
2-(3-Trifluoromethylanilino)nicotinic acid ≥95%
2-(3-Trifluoromethylanilino)nicotinic acid

Hazard: Irritant

Formula: C₁₃H₉F₃N₂O₂
MDL Number: MFCD00010569
CAS Number: 4394-00-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR