(S)-(+)-1-Aminopropan-2-ol ≥95%

Supplier: Matrix Scientific

076112-10G 076112-100G
102544-550EA 178.46 USD
102544-550 102544-552
(S)-(+)-1-Aminopropan-2-ol ≥95%
(S)-(+)-1-Aminopropan-2-ol

Hazard: Irritant

Formula: C₃H₉NO
MDL Number: MFCD00064429
CAS Number: 2799-17-9
UN: 3259
ADR: 8,II

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR