4-(4-Hydroxy-4-methylpentyl)cyclohex-3-enecarbaldehyde ≥95%

Supplier: Matrix Scientific

Synonyms: Hydroxyisohexyl 3-cyclohexene carboxaldehyde

076111-1G 076111-5G
102544-546EA 272.31 USD
102544-546 102544-548
4-(4-Hydroxy-4-methylpentyl)cyclohex-3-enecarbaldehyde ≥95%
4-(4-Hydroxy-4-methylpentyl)cyclohex-3-enecarbaldehyde

Hazard: Irritant

Formula: C₁₃H₂₂O₂
MW: 210.32 g/mol
MDL Number: MFCD00019423
CAS Number: 31906-04-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR