1-Hydroxycyclohexyl phenyl ketone ≥95%

Supplier: Matrix Scientific

Synonyms: (1-Hydroxycyclohexyl)(phenyl)methanone

076108-5G 076108-10G 076108-100G
102544-530EA 36.92 USD
102544-530 102544-532 102544-534
1-Hydroxycyclohexyl phenyl ketone ≥95%
1-Hydroxycyclohexyl phenyl ketone

TCA listed: T
Hazard: Irritant

Formula: C₁₃H₁₆O₂
MW: 204.27 g/mol
Boiling Pt: >225 °C
Melting Pt: 46…49 °C
Density: 1.17
Flash Pt: >150 °C
MDL Number: MFCD00059561
CAS Number: 947-19-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR