4-Chloro-2,3-dimethylpyridine-N-oxide ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Chloro-2,3-dimethylpyridine 1-oxide

076107-5G 076107-25G
102544-526EA 83.08 USD
102544-526 102552-012
4-Chloro-2,3-dimethylpyridine-N-oxide ≥95%
4-Chloro-2,3-dimethylpyridine-N-oxide

Hazard: Irritant-Harmful

Formula: C₇H₈ClNO
MW: 157.60 g/mol
Melting Pt: 104…106 °C
MDL Number: MFCD06411125
CAS Number: 59886-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR