2-Methoxycinnamaldehyde ≥95%
Supplier: Matrix Scientific
076106-5G
076106-10G
076106-100G
102544-518EA
364.62
USD
102544-518
102544-520
102544-522
2-Methoxycinnamaldehyde ≥95%
2-Methoxycinnamaldehyde
Hazard: Irritant
Formula:
C₁₀H₁₀O₂ MW: 162.1852 g/mol |
MDL Number:
MFCD00007001 CAS Number: 1504-74-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...