2,9-Dimethylquinolino[2,3-b]acridine-7,14(5H,12H)-dione ≥95%

Supplier: Matrix Scientific

076105-1G 076105-5G
102544-514EA 246.15 USD
102544-514 102544-516
2,9-Dimethylquinolino[2,3-b]acridine-7,14(5H,12H)-dione ≥95%
2,9-Dimethylquinolino[2,3-b]acridine-7,14(5H,12H)-dione

Hazard: Irritant

Formula: C₂₂H₁₆N₂O₂
MDL Number: MFCD00071968
CAS Number: 980-26-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR