7,7'-(Carbonylbis(azanediyl))bis(4-hydroxynaphthalene-2-sulfonic acid) ≥95%

Supplier: Matrix Scientific

076104-1G 076104-5G
102544-510EA 178.46 USD
102544-510 102544-512
7,7'-(Carbonylbis(azanediyl))bis(4-hydroxynaphthalene-2-sulfonic acid) ≥95%
7,7'-(Carbonylbis(azanediyl))bis(4-hydroxynaphthalene-2-sulfonic acid)

Hazard: Irritant

Formula: C₂₁H₁₆N₂O₉S₂
MDL Number: MFCD00035715
CAS Number: 134-47-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR