Methyl-3-chloro-4-fluorobenzoate ≥95%

Supplier: Matrix Scientific

Synonyms: Methyl 3-chloro-4-fluorobenzoate

076102-1G 076102-5G 076102-10G
102544-498EA 143.08 USD
102544-498 102544-500 102544-502
Methyl-3-chloro-4-fluorobenzoate ≥95%
Methyl-3-chloro-4-fluorobenzoate

Hazard: Irritant

Formula: C₈H₆ClFO₂
MW: 188.59 g/mol
MDL Number: MFCD06203675
CAS Number: 234082-35-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR