Manidipine dihydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-[4-(Diphenylmethyl)-1-piperazinyl]ethyl Methyl 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarb, 1,4-Dihydro-2,6-dimethyl-4-(3-nitrophenyl)-3,5-pyridinedicarboxylic acid 2-[4-(Diphenylmethyl)-1-piperazinyl]ethyl Methyl Ester Dihydrochloride, 3-(2-(4-Benzhydrylpiperazin-1-yl)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate dihydrochloride

076101-1G 076101-5G
102544-494EA 126.15 USD
102544-494 102544-496
Manidipine dihydrochloride ≥95%
Manidipine dihydrochloride

Hazard: Irritant

Formula: C₃₅H₃₈N₄O₆
MW: 683.63 g/mol
MDL Number: MFCD00896434
CAS Number: 89226-75-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR