ω-Bromo-3-methoxyacetophenone ≥95%
Supplier: Matrix Scientific
076099-5G
076099-10G
102544-484EA
150.77
USD
102544-484
102544-486
ω-Bromo-3-methoxyacetophenone ≥95%
ω-Bromo-3-methoxyacetophenone
Hazard: Irritant
Formula:
C₉H₉BrO₂ |
MDL Number:
MFCD00000199 CAS Number: 5000-65-7 UN: 3261 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...