5,5'-Dibromo-2,2'-bithiophene ≥95%

Supplier: Matrix Scientific

076098-1G 076098-5G 076098-10G
102544-478EA 107.69 USD
102544-478 102544-480 102544-482
5,5'-Dibromo-2,2'-bithiophene ≥95%
5,5'-Dibromo-2,2'-bithiophene

Hazard: Irritant

Formula: C₈H₄Br₂S₂
MW: 324.05 g/mol
MDL Number: MFCD00219110
CAS Number: 4805-22-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR