5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol) ≥95%

Supplier: Matrix Scientific

Synonyms: 5,5'-Diallylbiphenyl-2,2'-diol, 5,5'-Diallyl-[1,1'-biphenyl]-2,2'-diol, Magnolol

076095-1G 076095-5G
102544-462EA 284.62 USD
102544-462 102544-464
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol) ≥95%
5,5'-Diallyl-2,2'-dihydroxybiphenyl (Magnolol)

Hazard: Irritant

Formula: C₁₈H₁₈O₂
MW: 266.34 g/mol
MDL Number: MFCD00016658
CAS Number: 528-43-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR