Ranolazine ≥95%

Supplier: Matrix Scientific

Synonyms: N-(2,6-Dimethylphenyl)-2-(4-(2-hydroxy-3-(2-methoxyphenoxy)propyl)piperazin-1-yl)acetamide

076093-5G 076093-10G
102544-454EA 66.15 USD
102544-454 102544-456
Ranolazine ≥95%
Ranolazine

Hazard: Irritant

Formula: C₂₄H₃₃N₃O₄
MW: 427.545 g/mol
MDL Number: MFCD00864690
CAS Number: 95635-55-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR