Desvenlafaxine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-(Dimethylamino)-1-(1-hydroxycyclohexyl)-ethyl)phenol, O-Desmethylvenlafaxine, 4-[2-(Dimethylamino)-1-(1-hydroxycyclohexyl)ethyl]phenol, Desvenlafaxine

076092-1G 076092-5G
102544-448EA 76.92 USD
102544-448 102544-450
Desvenlafaxine ≥95%
Desvenlafaxine

Hazard: Irritant

Formula: C₁₆H₂₅NO₂
MW: 263.38 g/mol
MDL Number: MFCD00871934
CAS Number: 93413-62-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR