N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥95%

Supplier: Matrix Scientific

Synonyms: Gefitinib

076091-1G 076091-5G
102544-444EA 73.85 USD
102544-444 102544-446
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine ≥95%
N-(3-Chloro-4-fluorophenyl)-7-methoxy-6-(3-morpholinopropoxy)quinazolin-4-amine

Hazard: Irritant

Formula: C₂₂H₂₄ClFN₄O₃
MW: 446.91 g/mol
MDL Number: MFCD04307832
CAS Number: 184475-35-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR