Bromotrispyrrolidinophosphonium hexafluorophosphate ≥95%
Supplier: Matrix Scientific
Synonyms:
Bromotri(1-pyrrolidinyl)phosphonium hexafluorophosphate, PyBrOP , Bromotripyrrolidinylphosphonium hexafluorophosphate, Bromotri(pyrrolidin-1-yl)phosphonium hexafluorophosphate(V)
076089-100G
076089-25G
102544-438EA
407.69
USD
102544-438
102552-010
Bromotrispyrrolidinophosphonium hexafluorophosphate ≥95%
Bromo-tris-pyrrolidinophosphonium hexafluorophosphate
Hazard: Irritant
Formula:
C₁₂H₂₄BrF₆N₃P₂ MW: 466.2 g/mol |
MDL Number:
MFCD00077412 CAS Number: 132705-51-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...