(Z)-tert-Butyl-2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate ≥95%

Supplier: Matrix Scientific

Synonyms: (Z)-tert-Butyl 2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate

076087-1G 076087-5G 076087-10G
102544-426EA 107.69 USD
102544-426 102544-428 102544-430
(Z)-tert-Butyl-2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate ≥95%
(Z)-tert-Butyl-2-(((1-(2-aminothiazol-4-yl)-2-(benzo[d]thiazol-2-ylthio)-2-oxoethylidene)amino)oxy)-2-methylpropanoate

Hazard: Irritant

Formula: C₂₀H₂₂N₄O₄S₃
MW: 478.61 g/mol
MDL Number: MFCD00071548
CAS Number: 89604-92-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR