Ethyl-3-bromo-5-nitrobenzoate ≥95%
Supplier: Matrix Scientific
Synonyms:
Ethyl 3-bromo-5-nitrobenzoate, 3-Bromobenzoic acid ethyl ester
076086-1G
076086-5G
102544-422EA
178.46
USD
102544-422
102544-424
Ethyl-3-bromo-5-nitrobenzoate ≥95%
Ethyl-3-bromo-5-nitrobenzoate
Hazard: Irritant
Formula:
C₉H₈BrNO₄ MW: 274.07 g/mol |
MDL Number:
MFCD09258760 CAS Number: 690260-94-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...