Calcein ≥95%

Supplier: Matrix Scientific

Synonyms: Oftasceina, Fluorescein Complexon, Oftasceinum, 2,7-Bis[bis(carboxymethyl)aminomethyl]fluorescein, Oftasceine, 2,2',2'',2'''-(((3',6'-Dihydroxy-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-2',7'-diyl)bis(methylene))bis(azanetriyl))tetraacetic acid, Fluorexon, Bis[N,N-bis(carboxymethyl)aminomethyl]fluorescein, NSC 298193

076082-5G 076082-10G 076082-100G
102544-398EA 53.85 USD
102544-398 102544-400 102544-402
Calcein ≥95%
Calcein

Hazard: Irritant

Formula: C₃₀H₂₆N₂O₁₃
MDL Number: MFCD00005049
CAS Number: 1461-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR