(R)-2-Acetamidopentanedioic acid ≥95%

Supplier: Matrix Scientific

076076-1G 076076-5G 076076-10G
102544-370EA 107.69 USD
102544-370 102544-372 102544-374
(R)-2-Acetamidopentanedioic acid ≥95%
(R)-2-Acetamidopentanedioic acid

Hazard: Irritant

Formula: C₇H₁₁NO₅
MDL Number: MFCD00063196
CAS Number: 19146-55-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR