Gatifloxacin sesquihydrate ≥95%
Supplier: Matrix Scientific
Synonyms:
1-Cyclopropyl-6-fluoro-8-methoxy-7-(3-methylpiperazin-1-yl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid sesquihydrate
076075-1G
076075-5G
102544-366EA
233.85
USD
102544-366
102544-368
Gatifloxacin sesquihydrate ≥95%
Gatifloxacin sesquihydrate
Purity: 95+%
CAS: 180200-66-2
MDL Number: MFCD22373645
Molecular Formula: C38H50F2N6O11
Molecular Weight: 804,85
Hazard: Irritant
Formula:
C₃₈H₅₀F₂N₆O₁₁ MW: 804.83 g/mol |
MDL Number:
MFCD22373645 CAS Number: 180200-66-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...