(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene)diisopropyl dicarbonate ≥95%

Supplier: Matrix Scientific

Synonyms: (R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene) diisopropyl dicarbonate, Tenofovir Disoproxil

076070-1G 076070-5G
102544-344EA 100 USD
102544-344 102544-346
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene)diisopropyl dicarbonate ≥95%
(R)-(((((1-(6-Amino-9H-purin-9-yl)propan-2-yl)oxy)methyl)phosphoryl)bis(oxy))bis(methylene)diisopropyl dicarbonate

Hazard: Irritant

Formula: C₁₉H₃₀N₅O₁₀P
MW: 519.44 g/mol
MDL Number: MFCD00952920
CAS Number: 201341-05-1

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR