manidipine ≥95%

Supplier: Matrix Scientific

Synonyms: 3-(2-(4-Benzhydrylpiperazin-1-yl)ethyl) 5-methyl 2,6-dimethyl-4-(3-nitrophenyl)-1,4-dihydropyridine-3,5-dicarboxylate

076068-250MG 076068-1G 076068-5G
102544-332EA 498.46 USD
102544-332 102544-334 102544-336
manidipine ≥95%
manidipine

Hazard: Irritant

Formula: C₃₅H₃₈N₄O₆
MW: 610.711 g/mol
MDL Number: MFCD00871291
CAS Number: 89226-50-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR