Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate ≥95%

Supplier: Matrix Scientific

Synonyms: 2,2'-((5-Carboxy-4-(carboxymethyl)-3-cyanothiophen-2-yl)azanediyl)diacetic acid, distrontium salt, Strontium Ranelate

076067-1G 076067-5G
102544-328EA 15.38 USD
102544-328 102544-330
Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate ≥95%
Strontium-2,2'-((5-carboxylato-4-(carboxylatomethyl)-3-cyanothiophen-2-yl)azanediyl)diacetate

Hazard: Irritant/Store in Freezer

Formula: C₁₂H₆N₂O₈SSr₂
MDL Number: MFCD09038742
CAS Number: 135459-87-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR