Fexofenadine hydrochloride ≥95%

Supplier: Matrix Scientific

Synonyms: 2-(4-(1-Hydroxy-4-(4-(hydroxydiphenylmethyl)piperidin-1-yl)butyl)phenyl)-2-methylpropanoic acid hydrochloride, Fexofenadine HCl

076066-1G 076066-5G
102544-324EA 889.23 USD
102544-324 102544-326
Fexofenadine hydrochloride ≥95%
Fexofenadine hydrochloride

Hazard: Irritant

Formula: C₃₂H₃₉NO₄
MDL Number: MFCD00865710
CAS Number: 153439-40-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR