1-(3,5,5,6,8,8-Hexamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone ≥95%

Supplier: Matrix Scientific

Synonyms: Tonalid

076064-250MG 076064-1G 076064-5G
102544-314EA 400 USD
102544-314 102544-316 102544-318
1-(3,5,5,6,8,8-Hexamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone ≥95%
1-(3,5,5,6,8,8-Hexamethyl-5,6,7,8-tetrahydronaphthalen-2-yl)ethanone

Hazard: Irritant

Formula: C₁₈H₂₆O
MW: 258.41 g/mol
MDL Number: MFCD00437355
CAS Number: 21145-77-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR