Indinavir sulfate ≥95%

Supplier: Matrix Scientific

076062-1G 076062-5G
102544-306EA 1069.23 USD
102544-306 102544-308
Indinavir sulfate ≥95%
Indinavir sulfate

Hazard: Irritant

Formula: C₃₆H₄₉N₅O₈S
MDL Number: MFCD00920346
CAS Number: 157810-81-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR