Hydroxychloroquine sulfate ≥95%

Supplier: Matrix Scientific

076061-1G 076061-5G
102544-300EA 143.08 USD
102544-300 102544-302
Hydroxychloroquine sulfate ≥95%
Hydroxychloroquine sulfate

Hazard: Irritant

Formula: C₁₈H₂₆ClN₃O
MDL Number: MFCD00078203
CAS Number: 747-36-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR