(2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol ≥95%

Supplier: Matrix Scientific

Synonyms: 2-Fluoroadenine Arabinosid, (2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol, 9-beta-D-Arabinofuranosyl-2-fluoro-9H-purin-6-amine, 9-beta-D-Arabinofuranosyl-2-fluoroadenine, Fludarabine

076060-250MG 076060-1G 076060-5G
102544-294EA 498.46 USD
102544-294 102544-296 102544-298
(2R,3S,4S,5R)-2-(6-Amino-2-fluoro-9H-purin-9-yl)-5-(hydroxymethyl)tetrahydrofuran-3,4-diol ≥95%
Arabinofuranosyl-2-fluoroadenine

Hazard: Irritant

Formula: C₁₀H₁₂FN₅O₄
MW: 285.24 g/mol
MDL Number: MFCD00132942
CAS Number: 21679-14-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR