5-Methylpyrazine-2-carboxylic acid-4-oxide ≥95%

Supplier: Matrix Scientific

Synonyms: Acipimox, 5-Carboxy-2-methylpyrazine 1-oxide, 2-Carboxy-5-methylpyrazine 4-Oxide, 5-Carboxy-2-methylpyrazine 1-Oxide

076057-1G 076057-5G
102544-280EA 178.46 USD
102544-280 102544-282
5-Methylpyrazine-2-carboxylic acid-4-oxide ≥95%
5-Methylpyrazine-2-carboxylic acid-4-oxide

Hazard: Irritant

Formula: C₆H₆N₂O₃
MW: 154.13 g/mol
MDL Number: MFCD00865757
CAS Number: 51037-30-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR