Lafutidine ≥95%
Supplier: Matrix Scientific
|
Warning
|
Synonyms:
(+)-(Z)-2-((Furan-2-ylmethyl)sulphinyl)-N-(4-((4-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide, FRG-8813, (Z)-2-((Furan-2-ylmethyl)sulphinyl)-N-(4-((3-(piperidin-1-ylmethyl)pyridin-2-yl)oxy)but-2-en-1-yl)acetamide, (+)-Lafutidine
076055-250MG
076055-1G
102544-272EA
640
USD
102544-272
102544-274
Lafutidine ≥95%
Lafutidine
Purity: 95+%
CAS: 118288-08-7
MDL Number: MFCD21607521
Molecular Formula: C22H29N3O4S
Molecular Weight: 431,56
Hazard: Irritant
Formula:
C₂₂H₂₉N₃O₄S MW: 431.55 g/mol |
MDL Number:
MFCD21607521 CAS Number: 118288-08-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...