Pemetrexed ≥95%

Supplier: Matrix Scientific

Synonyms: (S)-2-(4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo-[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioic acid, (S)-2-(4-(2-(2-Amino-4-oxo-4,7-dihydro-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl)benzamido)pentanedioic acid, Alimta, [3H]-, N-[4-[2-(2-Amino-4,7-dihydro-4-oxo-1H-pyrrolo[2,3-d]pyrimidin-5-yl)ethyl]benzoyl]-L-glutamic Acid

076054-1G 076054-5G
102544-268EA 300 USD
102544-268 102544-270
Pemetrexed ≥95%
Pemetrexed

Hazard: Irritant

Formula: C₂₀H₂₁N₅O₆
MW: 427.42 g/mol
MDL Number: MFCD19105065
CAS Number: 137281-23-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR