(7a,17a)-17-Hydroxy-3-oxo-pregna-4,9(11)-diene-7,21-dicarboxylic acid g-lactone methyl ester ≥95%
Supplier: Matrix Scientific
Synonyms:
(7α,17α)-17-Hydroxy-3-oxo-pregna-4,9(11)-diene-7,21-dicarboxylic acid γ-Lactone 7-Methyl Ester, (7A,17A)-17-Hydroxy-3-oxo-pregna-4,9(11)-diene-7,21-dicarboxylic acid g-lactone methyl ester, 7α-(Methoxycarbonyl)-9(11)Δ-canrenone, Δ9,11-Eplerenone, Eplerenone EP Impurity C
076048-1G
076048-5G
102544-244EA
284.62
USD
102544-244
102544-246
(7a,17a)-17-Hydroxy-3-oxo-pregna-4,9(11)-diene-7,21-dicarboxylic acid g-lactone methyl ester ≥95%
(7a,17a)-17-Hydroxy-3-oxo-pregna-4,9(11)-diene-7,21-dicarboxylic acid g-lactone methyl ester
Hazard: Irritant
Formula:
C₂₄H₃₀O₅ |
MDL Number:
MFCD01244531 CAS Number: 95716-70-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...