Olmesartan medoxomil ≥95%

Supplier: Matrix Scientific

Synonyms: (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl 1-((2'-(2H-tetrazol-5-yl)-[1,1'-biphenyl]-4-yl)methyl)-4-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-5-carboxylate, (5-Methyl-2-oxo-1,3-dioxol-4-yl)methyl-1-((2'-(1H-tetrazol-5-yl)[1,1'-biphenyl]-4-yl)methyl)-4-(2-hydroxypropan-2-yl)-2-propyl-1H-imidazole-5-carboxylate

076046-1G 076046-5G
102544-238EA 107.69 USD
102544-238 102544-240
Olmesartan medoxomil ≥95%
Olmesartan medoxomil

Hazard: Irritant

Formula: C₂₉H₃₀N₆O₆
MW: 558.60 g/mol
MDL Number: MFCD00944911
CAS Number: 144689-63-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR