4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid ≥95%

Supplier: Matrix Scientific

Synonyms: Tamibarotene

076041-250MG 076041-1G
102544-220EA 967.69 USD
102544-220 102544-222
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid ≥95%
4-((5,5,8,8-Tetramethyl-5,6,7,8-tetrahydronaphthalen-2-yl)carbamoyl)benzoic acid

Hazard: Irritant

Formula: C₂₂H₂₅NO₃
MW: 351.45 g/mol
MDL Number: MFCD00866188
CAS Number: 94497-51-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR