N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)-benzofuran-5-yl)methanesulfonamide hydrochloride ≥95%

Supplier: Matrix Scientific

076039-250MG 076039-1G
102544-214EA 1138.46 USD
102544-214 102544-216
N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)-benzofuran-5-yl)methanesulfonamide hydrochloride ≥95%
N-(2-Butyl-3-(4-(3-(dibutylamino)propoxy)benzoyl)-benzofuran-5-yl)methanesulfonamide hydrochloride

Hazard: Irritant

Formula: C₃₁H₄₅ClN₂O₅S
MDL Number: MFCD00914940
CAS Number: 141625-93-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR