3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one ≥95%

Supplier: Matrix Scientific

076036-1G 076036-5G 076036-25G
102544-204EA 58.46 USD
102544-204 102552-002 102552-004
3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one ≥95%
3-(Chloromethyl)-1H-1,2,4-triazol-5(4H)-one

Hazard: Irritant

Formula: C₃H₄ClN₃O
MW: 133.54 g/mol
MDL Number: MFCD07776630
CAS Number: 252742-72-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR