Enzalutamide ≥95%
Supplier: Matrix Scientific
Synonyms:
4-(3-(4-Cyano-3-(trifluoromethyl)phenyl)-5,5-dimethyl-4-oxo-2-thioxoimidazolidin-1-yl)-2-fluoro-N-methylbenzamide, 4-[3-[4-Cyano-3-(trifluoromethyl)phenyl]-5,5-dimethyl-4-oxo-2-thioxo-imidazolidin-1-yl]-2-fluoro-N-methyl-benzamide, MDV-3100
076035-250MG
076035-1G
102544-198EA
946.15
USD
102544-198
102544-200
Enzalutamide ≥95%
Enzalutamide
Hazard: Irritant
Formula:
C₂₁H₁₆F₄N₄O₂S MW: 464.44 g/mol |
MDL Number:
MFCD14155804 CAS Number: 915087-33-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...