Ethyl-4-hydroxy-3-methoxycinnamate ≥95%

Supplier: Matrix Scientific

Synonyms: 4-Hydroxy-3-methoxycinnamic Acid Ethyl Ester, Ethyl 3-(4-hydroxy-3-methoxyphenyl)acrylate, Ethyl Ferulate, Ferulic Acid Ethyl Ester

076033-5G 076033-10G 076033-100G
102544-190EA 72.31 USD
102544-190 102544-192 102544-194
Ethyl-4-hydroxy-3-methoxycinnamate ≥95%
Ethyl-4-hydroxy-3-methoxycinnamate

Hazard: Irritant

Formula: C₁₂H₁₄O₄
MW: 222.24 g/mol
Boiling Pt: 164…166 °C (0.5 mmHg)
Melting Pt: 63…65 °C
MDL Number: MFCD00009190
CAS Number: 4046-02-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR