1,4-Diiodobenzene ≥95%

Supplier: Matrix Scientific

076032-5G 076032-10G 076032-100G
102544-184EA 90.77 USD
102544-184 102544-186 102544-188
1,4-Diiodobenzene ≥95%
1,4-Diiodobenzene

Hazard: Irritant

Formula: C₆H₄I₂
MW: 329.91 g/mol
Boiling Pt: 285
Melting Pt: 131…133 °C
MDL Number: MFCD00001054
CAS Number: 624-38-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR