3-Bromocatechol ≥95%

Supplier: Matrix Scientific

Synonyms: 3-Bromobenzene-1,2-diol, 1-Bromo-2,3-dihydroxybenzene

076031-1G 076031-5G
102544-180EA 106.15 USD
102544-180 102544-182
3-Bromocatechol ≥95%
3-Bromocatechol

Hazard: Irritant

Formula: C₆H₅BrO₂
MW: 189.01 g/mol
MDL Number: MFCD00869768
CAS Number: 14381-51-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR