N-(2-Hydroxyethyl)morpholine ≥95%

Supplier: Matrix Scientific

Synonyms: 4-(2-Hydroxyethyl)morpholine, 2-Morpholinoethanol

076025-10G 076025-100G
102544-154EA 29.23 USD
102544-154 102544-156
N-(2-Hydroxyethyl)morpholine ≥95%
N-(2-Hydroxyethyl)morpholine

Hazard: Irritant

Formula: C₆H₁₃NO₂
MW: 131.17 g/mol
MDL Number: MFCD00006180
CAS Number: 622-40-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR